Alfa Aesar™ Bismuth titanium isopropoxide, 5% w/v in isopropanol

Preparation of thin film mixed oxides of Ti and Bi

Manufacturer: Alfa Aesar™ 42839AE

Quantity 100mL

 View more versions of this product

Catalog No. AA42839AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 338391-61-0
    Molecular Formula C21H49BiO7Ti
    Linear Formula BiTi[OCH(CH3)2]7
    Formula Weight 670.48
    MDL Number MFCD01865878
    UN Number UN1219
    Chemical Name or Material Bismuth titanium isopropoxide

    Specifications

    Formula Weight 670.48
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration 5% w/v in isopropanol
    Linear Formula BiTi[OCH(CH3)2]7
    CAS 338391-61-0
    Storage Note 1
    • Packaged under Argon
    Quantity 100mL
    Physical Form Liquid
    Molecular Formula C21H49BiO7Ti
    MDL Number MFCD01865878
    UN Number UN1219
    Color Pale yellow
    Sensitivity Moisture Sensitive
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Chemical Name or Material Bismuth titanium isopropoxide

    GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit