Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Tribromophenate, Powder, 40-49%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5175-83-7
5175-83-7
CAS Max % 1
Arsenic (As) 2e-06
Percent Purity 40 to 49%
Grade Reagent
Copper (Cu) To Pass Test
Lead (Pb) To Pass Test
Loss on Drying 0.03
Packaging Poly Pail
Silver (Ag) To Pass Test
Sulfate To Pass Test
Quantity 1 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.