Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Tribromophenate, Powder, 40-49%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5175-83-7
5175-83-7
Grade Reagent
CAS Max % 1
Arsenic (As) 2e-06
Percent Purity 40 to 49%
Copper (Cu) To Pass Test
Lead (Pb) To Pass Test
Loss on Drying 0.03
Packaging Fiber Drum
Silver (Ag) To Pass Test
Sulfate To Pass Test
Quantity 50 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.