Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth tris(2,2,6,6-tetramethyl-3,5-heptanedionate), 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
142617-53-6
C33H57BiO6
758.793
MFCD00064763
ZNHBPQSSVCRFST-LWTKGLMZSA-K
tris 2,2,6,6-tetramethyl-3,5-heptanedionato bismuth iii,bismuth iii tetramethylheptanedionate,4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy bismuthanyl oxy-2,2,6,6-tetramethylhept-4-en-3-one
16717622
(Z)-5-bis[[(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl]oxy]bismuthanyloxy-2,2,6,6-tetramethylhept-4-en-3-one
CC(C)(C)C(=CC(=O)C(C)(C)C)O[Bi](OC(=CC(=O)C(C)(C)C)C(C)(C)C)OC(=CC(=O)C(C)(C)C)C(C)(C)C
142617-53-6
C33H57BiO6
758.793
MFCD00064763
ZNHBPQSSVCRFST-LWTKGLMZSA-K
tris 2,2,6,6-tetramethyl-3,5-heptanedionato bismuth iii,bismuth iii tetramethylheptanedionate,4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy bismuthanyl oxy-2,2,6,6-tetramethylhept-4-en-3-one
16717622
(Z)-5-bis[[(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl]oxy]bismuthanyloxy-2,2,6,6-tetramethylhept-4-en-3-one
CC(C)(C)C(=CC(=O)C(C)(C)C)O[Bi](OC(=CC(=O)C(C)(C)C)C(C)(C)C)OC(=CC(=O)C(C)(C)C)C(C)(C)C
Infrared Spectrum Authentic
Color White to Yellow
Packaging Glass bottle
Quantity 1g
Assay Percent Range 97%
Formula Weight 758.79
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Bismuth tris(2,2,6,6-tetramethyl-3,5-heptanedionate)

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.