Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bisoprolol Fumarate, USP, 97.5-102%, Spectrum™

Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail

B2497, 104344-23-2, (C18H31NO4)2•C4H4O4

Spectrum™ Bisoprolol Fumarate, USP
Packaging Poly Pail
Ignition Residue 0.001
Water 0.005
Specific Rotation +2°
Quantity 5 kg
CAS 104344-23-2
CAS Max % 1
Molecular Formula C40H66N2O12
InChI Key VMDFASMUILANOL-WXXKFALUNA-N
SMILES OC(=O)\C=C\C(O)=O.CC(C)NCC(O)COC1=CC=C(COCCOC(C)C)C=C1.CC(C)NCC(O)COC1=CC=C(COCCOC(C)C)C=C1
IUPAC Name (2E)-but-2-enedioic acid; bis(1-[(propan-2-yl)amino]-3-(4-{[2-(propan-2-yloxy)ethoxy]methyl}phenoxy)propan-2-ol)
Molecular Weight (g/mol) 766.97
Percent Purity 97.5 to 102%
Show More Show Less