Alfa Aesar™ Bisoprolol hemifumarate salt, 98%

Manufacturer: Alfa Aesar™ H56014MC

Quantity 100mg

 View more versions of this product

Catalog No. AAH56014MC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 104344-23-2
    Molecular Formula C18H31NO4·0.5C4H4O4
    Formula Weight 383.49
    MDL Number MFCD00792743
    Merck Index 14,1295
    Chemical Name or Material Bisoprolol hemifumarate salt

    Specifications

    Formula Weight 383.49
    Molecular Formula C18H31NO4·0.5C4H4O4
    MDL Number MFCD00792743
    CAS 104344-23-2
    Merck Index 14,1295
    Quantity 100mg
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number UB8390000
    Chemical Name or Material Bisoprolol hemifumarate salt
    Assay 98%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit