Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BisphenolA (2,3-dihydroxypropyl) glycidyl ether, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Bisphenol A (2,3-dihydroxypropyl) glycidyl ether can be obtained as a product of the reaction of bisphenol A with epichlorohydrin.It is generally used as a substrate to prepare epoxy resins, which is used in coatings and adhesives.

$146.00 - $444.50

Specifications

Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Linear Formula C21H26O5
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-5445
View Documents
MilliporeSigma Supelco
7341725MG
25 mg
Each for $146.00
Add to cart
 
11-101-5444
View Documents
MilliporeSigma Supelco
73417100MG
100 mg
Each for $444.50
Add to cart
 
Description

Description

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

≥95.0% (HPLC)
Analytical Standard
C21H26O5
Neat
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit