Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BisphenolA (3-chloro-2-hydroxypropyl) glycidyl ether, ≥90% (HPLC), MilliporeSigma™ Supelco™

Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether is a chlorinated derivative, which is released during the thermal coating treatment of food cans, big storage vessels and food containers.

$325.00

Specifications

Percent Purity ≥90% (HPLC)
Grade Analytical Standard
Linear Formula C21H25 ClO4
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-2340
View Documents
MilliporeSigma Supelco
7312425MG
25 mg
Each for $99.70
N/A
Due to product restrictions, please to purchase or view availability for this product.
11-101-3025
View Documents
MilliporeSigma Supelco
73124100MG
100 mg
Each for $325.00
Add to cart
 
Description

Description

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

≥90% (HPLC)
Analytical Standard
C21H25 ClO4
Neat
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit