Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bisphosphoglycerate mutase Polyclonal Antibody
Bisphosphoglycerate mutase Polyclonal Antibody
Supplier: Thermo Scientific PA521821
Description
Bisphosphoglycerate mutase Polyclonal Antibody for Western Blot, ICC/IF

Specifications
Bisphosphoglycerate mutase | |
Polyclonal | |
Unconjugated | |
BPGM | |
2,3-bisphosphoglycerate mutase; 2,3-bisphosphoglycerate mutase, erythrocyte; 2,3-bisphosphoglycerate synthase; 2,3-diphosphoglycerate mutase; Ab2-098; AI323730; AL022789; Bisphosphoglycerate mutase; BPG-dependent PGAM; Bpgm; C86192; DPGM; erythrocyte 2,3-bisphosphoglycerate mutase; hypothetical protein LOC533785; testis secretory sperm-binding protein Li 202a; zgc:92230 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12183, 669 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.84 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P07738, P15327 | |
BPGM | |
Recombinant fragment corresponding to a region within amino acids 1 and 246 of BPGM (Uniprot ID#P07738). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction