Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biuret Reagent TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle

B-288

Spectrum™ Biuret Reagent TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS 1310-73-2,6381-59-5
CAS Max % 0.001,0.999
Molecular Formula HNaO
UN Number UN1824
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Packaging Amber Glass Bottle
Quantity 1 L