Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BIVM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562140
Description
Immunogen sequence: RSNDSGNGEH KSERKSPEEN LQGAVKSFCT SASGAPLGPK GDGHYPWSCP VTHTREKIYA ICSDYAFLNQ ATSIYKTPNP SRSPCLPDST SLSAGNNSSR Y Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 75%.
BIVM has been identified using an electronic search based on the conservation of short sequence motifs within the variable region of immunoglobulin (Ig) genes. It is tightly linked (41 bp) and in the opposite transcriptional orientation to MGC5302 (also known as KDEL1 and EP58) in human. BIVM has two isoforms with MW 57 kDa and 32 kDa.
Specifications
BIVM | |
Polyclonal | |
Unconjugated | |
BIVM | |
AI854267; basic immunoglobulin-like variable motif-containing protein; basic immunoglobulin-like variable motif-containing protein-like; basic, immunoglobulin-like variable motif containing; BIVM; LOC102548228; RP11-484I6.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UB2 | |
BIVM | |
Recombinant protein corresponding to Human BIVM. Recombinant protein control fragment (Product #RP-97956). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction