Tocris™ BIX 02189

Selective MEK5 and ERK5 inhibitor

Manufacturer: Tocris™ 4842/50

 View more versions of this product

Catalog No. 48-425-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors BIX 02189
    Purity >98
    Molecular Weight 440.54
    Quantity 50mg
    Product Type BIX 02189

    Selective MEK5 and ERK5 inhibitor (IC50 values are 1.5 and 59nM respectively). Inhibits sorbitol-induced ERK5 phosphorylation in HeLa cells. Selective for MEK5 and ERK5 over MEK1/2, ERK1/2 and a panel of 87 kinases. Induces apoptosis in acute myeloid leukemia tumor cells.

    • Chemical Name: (3Z)-3-[[[3-[(Dimethylamino)methyl]phenyl]amino]phenylmethylene]-2,3-dihydro-N,N-dimethyl-2-oxo-1H-indole-6-carboxamide
    • Formula: C27H28N4O2
    • CAS Number: 1265916-41-3
    • Solubility: Soluble to 100mM in DMSO and to 100mM in 1eq. HCl
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit