Learn More
BKV VP1 Mouse, Clone: 3G2, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB3204M23A
Catalog No. 89-032-134
Add to cart
Description
Description
Mouse monoclonal antibody raised against a full-length recombinant BKV VP1.
  • High specificity
  • High reactivity
Specifications
Specifications
BKV VP1
3G2
Mouse monoclonal antibody raised against a full-length recombinant BKV VP1.
NC_001538
VP1
Mouse
IgG1 Kappa
RUO
Primary
1489515
ELISA, Western Blot
Unconjugated
In ascites fluid
Ascites
BKPyVgp4
BKV VP1 (YP_717939.1, 1 a.a. ~ 369 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
200 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)