Learn More
Brady™ Black 6100 Series Thermal Transfer Printer Ribbons
Used for general purpose. Brady™ Black 6100 Series Thermal Transfer Printer Ribbons are available in various sizes.
Manufacturer:  Brady™ R6113
 View more versions of this product
Catalog No. 19-104-640
Add to cart
Description
Description
  • Fiber ribbon core
  • Wax formulation
Specifications
Specifications
Black
242 ft.
6100 Series Thermal Transfer Printer Ribbon
Ribbon Core Inside Diameter: 0.5 in.
4 in.