Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Sklar InstrumentsSupplier Diversity Partner Forceps, Sklar Black Adson Tissue Forceps, Sklar 4-3/4 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel with Reduced Reflectivity Polymer Coating, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 901060
Forceps, Sklar Black Adson Tissue Forceps, Sklar, 4-3/4 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel with Reduced Reflectivity Polymer Coating, EA. Sklar Black Adson Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.
Catalog No. 50-112-1770