Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Graves Vaginal Speculum, Sklar, Straight, Stainless Steel with Reduced Reflectivity Polymer Coating, Small With Loop To Hold and Direct Disposable Smoke Evacuation Tubing. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 909000
Sklar Black Graves Vaginal Speculum, Sklar, Straight, Stainless Steel with Reduced Reflectivity Polymer Coating, Small With Loop To Hold and Direct Disposable Smoke Evacuation Tubing. The Sklar Black Graves Vaginal Speculum is used for laser gynecological procedures. It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient. The Sklar Black Graves Vaginal Speculum is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.
Catalog No. 50-143-6897