Learn More
Fisherbrand™ Black Ribbed Phenolic Cap with PE Cone Liner Available on GSA/VA Contract for Federal Government customers only.
Forms exceptionally tight seal, offers good chemical barrier
 View more versions of this product
Catalog No. 02-911-727A
  Check Availability
Add to cart
Description
Description
  • Form tight seal
  • Offers good chemical barrier
  • Used to seal liquids, soaps, macro-invertebrates
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Phenolic
Black
PE
Polycone cap
Black Ribbed
Screw
Polycone
Sealing
1/PK.
Yes
38-400