Learn More
Fisherbrand™ Black Smooth Sided Mineral Filled Phenolic Cap with Rubber Liner
 View more versions of this product
Catalog No. 02-912-048A
  Check Availability
Add to cart
Description
Description
  • Liner is compliant with FDA 21 CFR 177.2600 Rubber Articles Intended for Repeated Use
  • Liner exhibit good resiliency and resistance to moisture vapor
  • Closures provide consistent adherence to close dimensional tolerance
Specifications
Specifications
Phenolic
PhenolicþPhenolic
576 Each/Cs.
Black
22-400
Screw
Black
Packed in 48 bags of 12
Rubber Lined Cap
Rubber
Round