Learn More
Fisherbrand™ Black Smooth Sided Mineral Filled Phenolic Cap with Rubber Liner Available on GSA/VA Contract for Federal Government customers only.
 View more versions of this product
Catalog No. 02-912-052A
  Check Availability
Add to cart
Description
Description
  • Liner is compliant with FDA 21 CFR 177.2600 Rubber Articles Intended for Repeated Use
  • Liner exhibit good resiliency and resistance to moisture vapor
  • Closures provide consistent adherence to close dimensional tolerance
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Phenolic
PhenolicþPhenolic
576 Each/Cs.
Black
38-400
Screw
Black
Packed in 48 bags of 12
Rubber Lined Cap
Rubber
Round