Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Pederson Vaginal Speculum, Sklar, 4 3/4 in. x 1 in. Sklar Black Pederson Vaginal Speculum, Large, With Permanent Smoke Evacuation Tube, Stainless Steel with Reduced Reflectivity Polymer Coating. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 909044
Sklar Black Pederson Vaginal Speculum, Sklar, 4 3/4 in. x 1 in. Sklar Black Pederson Vaginal Speculum, Large, With Permanent Smoke Evacuation Tube, Stainless Steel with Reduced Reflectivity Polymer Coating. The Sklar Black Pederson Vaginal Speculum is used for laser gynecological procedures. It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The blades of the Pederson Speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient. The Sklar Black Pederson Vaginal Speculum is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.
Catalog No. 50-143-6912