Learn More
Black Stallion Flame-Resistant Coat
$11.66 - $26.50
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-999-512
|
Black Stallion
F930CMD |
Medium |
Each for $11.66
|
|
19-088-1709
|
Black Stallion
F930CXL |
X-Large |
Each for $24.00
|
|
19-999-515
|
Black Stallion
F930C2X |
2X-Large |
Each for $25.50
|
|
19-999-516A
|
Black Stallion
F930C3X |
3X-Large |
Each for $26.50
|
|
Description
Flame-resistant treated cotton protects against heat and flames. Ideal for welding and other applications with potential for sparking. Green.Pants feature two side pockets, one rear pocket, and flame-retardant self-grip fasteners on waist and fly. 32 in. (81cm) inseam. Standard belt loops.
Jacket is 30 in. (76cm) long and has a snap front with a large inside pocket. Wrists are adjustable with snaps.
Specifications
Cotton | |
FR Coat |