Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Steiner-Auvard Weighted Speculum, Sklar, Permanent Smoke Evacuation Tube, 5-1/2 in. x 1-1/4 in., Blunt, Straight, Smooth, Extra Long Blade, Stainless Steel with Reduced Reflectivity Polymer Coating. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 903562
Sklar Black Steiner-Auvard Weighted Speculum, Sklar, Permanent Smoke Evacuation Tube, 5-1/2 in. x 1-1/4 in., Blunt, Straight, Smooth, Extra Long Blade, Stainless Steel with Reduced Reflectivity Polymer Coating. The Sklar Black Steiner-Auvard Weighted Vaginal Speculum is a single end vaginal speculum commonly used in laser OB/GYN procedures such as vaginal hysterectomy or dilation and curettage. This speculum features extra long blades, measuring 5-1/2 in. x 1-1/4 in. It is weighted with a medium, 2-1/2 lb. weight. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Catalog No. 50-143-6654