Learn More
Thermo Scientific™ Blade Holder Packages for the Cryotome™ FSE Cryostat Available on GSA/VA Contract for Federal Government customers only.
Hold blades for the Cryotome™ FSE Cryostat with these packages.
Manufacturer:  Thermo Scientific™ A78910033
 View more versions of this product
Catalog No. A78910033
  Check Availability
Add to cart
Specifications
Description
Hold blades for the Cryotome™ FSE Cryostat with these Thermo Scientific™ blade holder packages.
Universal Blade Holder:
  • Accepts standard C- or D-profile knife up to 220mm; 120mm is standard length
  • Use with standard steel knives or with disposable blade holder
  • Use in conjunction with disposable blade holders (Part Nos. 40 or 47)
Low-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder for use with low-profile disposable blades
High-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder will accept either standard or heavy-duty high-profile blades, providing a wide range of tissue sectioning options
Description & Specifications
Specifications
High-profile Cryotome Blade Holder
High-profile Cryotome Blade Holder
High profile cryotome blade holder
FSE/FE and SME/E
FSE/FE and SME/E