Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLAST-2 (CD23) Mouse anti-Human, Unconjugated, Clone: EBVCS3, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen BLAST-2 (CD23)
Applications Flow Cytometry, Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF23181MI
View Documents
MilliporeSigma
MABF2318100UG
100 μg
Each for $386.00
Add to cart
 
MABF23182MI
View Documents
MilliporeSigma
MABF231825UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

BLAST-2 (CD23) Monoclonal antibody specifically detects BLAST-2 (CD23) in Human samples. It is validated for Activity Assay, Flow Cytometry, Immunofluorescence.
Specifications

Specifications

BLAST-2 (CD23)
Unconjugated
Mouse
Low affinity immunoglobulin epsilon Fc receptor;C-type lectin domain family 4 member J;CD23;Fc-epsilon-RII;Immunoglobulin E-binding factor;Lymphocyte IgE receptor
EBV-transformed human cells.
Primary
Stable for 1 year at -20°C from date of receipt.
Flow Cytometry, Immunofluorescence
Purified
Human
FCER2, CD23A, CLEC4J, FCE2, IGEBF
Monoclonal
Ligatrap
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit