Learn More
Thermo Scientific Chemicals Blasticidin S hydrochloride, 10 mg/ml in HEPES buffer, sterile-filtered
Description
Blasticidin S hydrochloride, acts quickly by inhibiting peptide bond formation by the ribosome. The blasticidin S resistance gene (BSR), found in Bacillus cereus K55-S1, has been used in multiple studies for its use as a selective compound. Resistance is conferred by expression of one of two Blasticidin S deaminase genes: BSD from Aspergillus terreus or bsr from Bacillus cereus. These deaminases convert Blasticidin S to a nontoxic deaminohydroxy derivative. Blasticidin S is used for the prevention of contamination of cell cultures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Blasticidin S hydrochloride is an antibiotic that works by inhibiting peptide bond formation by the ribosome
Application
• Blasticidin S hydrochloride is a nucleoside antibiotic that inhibits protein synthesis in prokaryotic and eukaryotic cells
• It inhibits peptide bond formation by the ribosome
Specifications
Type | Blasticidin S Hydrochloride |
Physical Form | Liquid |
Solubility Information | Soluble in water. Insoluble in organic solvents. |
SMILES | CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl |
IUPAC Name | (2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride |
PubChem CID | 124080960 |
Concentration | 10mg/mL in HEPES buffer |
Quantity | 5 x 1 mL |
Chemical Name or Material | Blasticidin S hydrochloride, sterile-filtered |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
Precautionary Statement | GHS P Statement P264-P270-P301+P312-P330-P501a Wash thoroughly after handling. Do not eat, drink or smoke when using this product. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth. Dispose of contents/container in accordance with local/regional/national/international regulations. |
TSCA | No |
storageNote1 | −30° to −10°C |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.