Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Blasticidin S hydrochloride, ≥98%,/ml in 1M HEPES buffer soln.

Click to view available options
Quantity:
20mg
5 x 20mg
3513-03-9
C17H27ClN8O5
458.90
MFCD02091640
YQXYQOXRCNEATG-UHFFFAOYNA-N
124080960
(2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride
CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl
3513-03-9
C17H27ClN8O5
458.90
MFCD02091640
YQXYQOXRCNEATG-UHFFFAOYNA-N
124080960
(2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride
CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl

Inhibits protein synthesis and induces apoptosis

Applications
Blasticidin S hydrochloride is used to inhibit protein synthesis in prokaryotic and eukaryotic organisms. It also induces apoptosis.

Solubility
Soluble in water.

Notes
Store in cool place. Recommended to prepare and use solutions on the same day. Incompatible with strong oxidizing agents and strong bases.
UN Number UN2810
Merck Index 14,1316
Formula Weight 458.9
Physical Form Liquid
Percent Purity ≥98%
Color Colorless
Quantity 5 x 20mg
Chemical Name or Material Blasticidin S hydrochloride, 10mg/mL in 1M HEPES buffer soln.

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

storageNote1 : -30° to -10°C