Learn More
Selleck Chemical LLCSupplier Diversity Partner Bleomycin Sulfate 50mg 9041-93-4 NSC125066  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S121450MG
Bleomycin Sulfate (NSC125066) is a glycopeptide antibiotic and an anticancer agent for squamous cell carcinomas (SCC) with IC50 of 4 nM in UT-SCC-19A cells.
Catalog No. 50-673-5