Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BLID Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577185
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a BH3-like motif containing protein involved in cell death. The encoded protein may induce apoptosis in a caspase-dependent manner. The protein is localized in both the cytoplasm and the mitochondrion.
Specifications
BLID | |
Polyclonal | |
Unconjugated | |
BLID | |
BH3-like motif containing, cell death inducer; BH3-like motif-containing cell death inducer; BLID; BRCC2; breast cancer cell 2; breast cancer cell protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
414899 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZY5 | |
BLID | |
Recombinant full length Human BLID. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction