Learn More
at Anti-Blimp-1 Rat, Unlabeled, Clone: 6D3, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 564703
Description
The 6D3 monoclonal antibody specifically binds to mouse B lymphocyte-induced maturation protein 1 (Blimp-1) and crossreacts with human BLIMP-1. Blimp-1 is a 98 kDa zinc finger-containing protein that is encoded by the Prdm1 gene. Blimp-1 was first discovered as a transcriptional repressor of the IFN- β promoter. Blimp-1 is critical for primordial germ cells specification in the early embryo and induction of terminal cell differentiation in multiple somatic cell types. It is largely known as a master regulator of antibody-secreting plasma cells due to its function in the repression of PAX5 , CIITA and cMYC genes. Blimp-1 also controls T cell homeostasis and effector differentiation through different mechanisms, including repression of IL-17 producing cells. Blimp-1 also plays roles in the maturation and activity of dendritic cells and natural killer cells.
Specifications
at Anti-Blimp-1 | |
Monoclonal | |
0.5mg/mL | |
BLIMP-1; PRDM1; PR domain containing 1, with ZNF domain; PRDI-BF1 | |
Rat | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG2a κ |
Flow Cytometry | |
6D3 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Blimp-1 Recombinant Protein | |
0.1 mg | |
Primary | |
Store undiluted at 4°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.