Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BLOC1S3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106958
Description
Sequence homology: Cow: 93%; Dog: 86%; Goat: 79%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 100%; Rat: 100%; Sheep: 79%; Zebrafish: 79%.
Component of the BLOC-1 complex, a complex that is required for normal biogenesis of lysosome-related organelles (LRO), such as platelet dense granules and melanosomes. In concert with the AP-3 complex, the BLOC-1 complex is required to target membrane protein cargos into vesicles assembled at cell bodies for delivery into neurites and nerve terminals. The BLOC-1 complex, in association with SNARE proteins, is also proposed to be involved in neurite extension. Plays a role in intracellular vesicle trafficking.
Specifications
BLOC1S3 | |
Polyclonal | |
Unconjugated | |
BLOC1S3 | |
biogenesis of lysosomal organelles complex 1 subunit 3; biogenesis of lysosomal organelles complex-1, subunit 3; biogenesis of lysosome-related organelles complex 1 subunit 3; biogenesis of lysosome-related organelles complex-1, subunit 3; BLOC-1 subunit 3; BLOC1S3; BLOS3; HPS8; reduced pigmentation, mouse, homolog of; RP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
388552 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q6QNY0 | |
BLOC1S3 | |
Synthetic peptide located within the following region: RGRPTGLRVA GEAAETDSEP EPEPEPTAAP RDLPPLVVQR ESAEEAWGTE. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction