Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Block, 28 x 2.0 mL, EMD Products for Science Education

For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)

Manufacturer:  Thermo Scientific™ 88870104/EMD

Catalog No. S37343


Add to cart

Description

Description

Specifications

Specifications

28 x 2.0mL block
Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Accessory, Block
76 x 124 x 39 mm (3 x 4.9 x 1.5 in.)
28 x 2.0mL
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit