Learn More
Thermo Scientific™ Blocker™ BSA Blocking Buffers Available on GSA/VA Contract for Federal Government customers only.
Block membranes, plates and other samples with these ready-to-use 10X PBS or TBS solutions of bovine serum albumin for Western blot, ELISA and other detection methods.
$171.60 - $185.00
Specifications
Applications ELISA, IHC, WB
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
PI37520 Available on GSA/VA Contract for Federal Government customers only. View Documents Thermo Scientific™
37520
Blocker BSA in TBS (10X) Pack for $171.60
Add to cart
PI37525 Available on GSA/VA Contract for Federal Government customers only. View Documents Thermo Scientific™
37525
Blocker BSA in PBS (10X) Pack for $185.00
Add to cart
Description
Description
Thermo Scientific™ Blocker BSA Blocking Buffers are ready-to-use, 10X PBS or TBS solutions of bovine serum albumin protein for blocking steps in Western blotting, ELISA, immunohistochemistry and nucleic acid detection methods.
These blocking buffers are 10% (w/v) solutions of high-quality BSA that are useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. Blocker BSA Buffers are most frequently diluted 10-fold (to 1% BSA) for initial testing, but other buffer concentrations can be beneficial for specific systems. Blocker BSA is usually more effective than nonfat milk blocking buffers for biotin-avidin systems because it contains a single purified protein that is devoid endogenous biotin.
Highlights:
  • Purified protein – 10% solutions of high-quality bovine serum albumin; a single purified protein provides fewer chances of cross-reaction with assay components than serum or milk solutions
  • Convenient – concentrated formulation saves storage space and can be diluted easily to obtain optimal blocking results for specific applications
  • Easy to use – no waiting for powder to dissolve with this ready-to-dilute liquid concentrate
  • Flexible – available in PBS and TBS formulations to suit a variety of applications
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ELISA, IHC, WB