Learn More
LI-CORSupplier Diversity Partner ODYSSEY BLOCKER PBS 500ML DFS Item
Manufacturer:  LI-CORSupplier Diversity Partner 92740000
Odyssey Blocking Buffer (PBS), 500 mL. Non-mammalian Odyssey Blocker. This Blocking Reagent is in PBS (phosphate-buffered saline) containing 0.1% sodium azide.
Catalog No. NC9877369
  Check Availability
Add to cart
For SAVINGS of $73.55, you may also be interested in considering Thermo Scientific™ Pierce™ Protein-Free Blocking Buffers as an alternative option to catalog number NC9877369
Show Details
PI37572
PBS Formulation; 1L
  • Save $73.55 on Each