Learn More
Microbiologics™ Blood Culture Identification (BCID) Control Panel (Inactivated)
SDP
Microbiologics™ Blood Culture Identification (BCID) Control Panel (Inactivated) includes six inactivated microorganism pellets each comprised of 22 organisms.
Supplier:  Microbiologics™ 8215
Catalog No. 23-046-869
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
  • Acinetobacter baumannii derived from NCIMB 12457
  • Candida albicans derived from NCYC 1363
  • Candida glabrata derived from NCPF 8018
  • Candida krusei derived from NCPF 3100
  • Candida parapsilosis derived from NCPF 3207
  • Candida tropicalis derived from NCPF 8759
  • Enterobacter cloacae derived from NCTC 13464
  • Enterococcus faecalis (vanB) derived from NCTC 13379
  • Escherichia coli derived from NCTC 13476
  • Haemophilus influenzae derived from NCTC 13377
  • Klebsiella oxytoca derived from NCTC 11686
  • Klebsiella pneumoniae (KPC) derived from NCTC  13438
  • Listeria monocytogenes serovar 7 derived from NCTC 10890
  • Neisseria meningitidis derived from NCTC 10026
  • Proteus vulgaris derived from NCTC 4636
  • Pseudomonas aeruginosa derived from NCTC 10662
  • Serratia marcescens derived from NCTC 9743
  • Staphylococcus aureus (MRSA) derived from NCTC 12493
  • Staphylococcus epidermidis (MSSE) derived from NCIMB 8853
  • Streptococcus agalactiae derived from NCTC 8017
  • Streptococcus pneumoniae derived from NCIMB 13286
  • Streptococcus pyogenes derived from NCIMB 13285
Specifications
Specifications
IVD, CE Marked
Helix Elite
Helix Elite Control Reference Panels and Standards
Plastic Box
Room Temperature
United States
In vitro diagnostics
6
Safety and Handling
Safety and Handling
ShelfLife : 8 Months (Minimum)