Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blood Group Antigen H (O) Type 1 Mouse anti-Human, Clone: 17-206, eBioscience™ Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 14981095

Encompass

 View more versions of this product

Catalog No. 14-981-095


Add to cart

Description

Description

The MAb 17-206 recognizes the human blood group antigen H (O) type 1, a subtype primarily found in epithelial and secretory cells. The precursor H antigen is modified by addition of saccharides to carbohydrate chains of proteins and lipids on erythrocytes and epithelial cells. The type, attachment, location, and sequence of saccharides determines the blood group antigen subtype and is mediated by multiple glycosyl transferases. Depending on ABO blood type, the H antigen is converted into either the A antigen, B antigen, both the A and B antigen, or left unconverted (O antigen). Evidence that the blood group antigen H is not expressed in tumor cells and that the level correlates with disease progression suggests that the blood group antigen H may be involved in cell proliferation, cell adhesion, and angiogenesis, although its exact function remains to be determined. The 17-206 antibody does not recognize blood group antigen H (O) type 2.

Blood Group Antigen H (O) Type 1 Monoclonal antibody specifically detects Blood Group Antigen H (O) Type 1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

Blood Group Antigen H (O) Type 1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Mouse
10 mg
Primary
4° C
Liquid
ELISA, Immunohistochemistry (Paraffin)
17-206
Unconjugated
ABH; BG-H; BG-H1; group h
Affinity chromatography
RUO
Human
Antibody
IgG3
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit