Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Blood Group B Transferase/GTB/ABO Rabbit anti-Human, Mouse, Clone: 2V4R0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Blood Group B Transferase/GTB/ABO |
---|---|
Clone | 2V4R0 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Blood Group B Transferase/GTB/ABO Monoclonal antibody specifically detects Blood Group B Transferase/GTB/ABO in Human, Mouse samples. It is validated for Western BlotSpecifications
Blood Group B Transferase/GTB/ABO | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
28 | |
IgG | |
Affinity purified |
2V4R0 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Blood Group B Transferase/GTB/ABO (P16442). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title