Learn More
Description
19-OLE reacts with H type 2 antigen, the basis of the ABO blood group system, involving three carbohydrate antigens: A, B, and H. A, B, and AB individuals express a glycosyltransferase activity that converts the H antigen to the A antigen (by addition of UDP-GalNAc) or to the B antigen (by addition of UDP-Gal), whereas O individuals lack such activity. It is expressed on endothelial cells, epithelial cells and granulocytes. Increased expression of this antigen has been observed on some tumor tissues such as gastric carcinomas, urothelial carcinomas, and colon carcinomas.
Specifications
Specifications
| Antigen | Blood Group H Type 2 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 19-OLE |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ABO |
| Gene Accession No. | P16442 |
| Gene Alias | ABH; BG-H; BG-H1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
