Learn More
BloodGroupAntigenB Mouse anti-Human, Clone: HEB-20, Abnova™
Mouse monoclonal antibody raised against native human Blood Group Antigen B.
Manufacturer:  Abnova Corporation MAB13296
Catalog No. 89-933-651
Add to cart
Description
Description
Specifications
Specifications
BloodGroupAntigenB
HEB-20
Mouse monoclonal antibody raised against native human Blood Group Antigen B.
In 10mM PBS.
Mouse
IgM, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Agglutination, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Agglutination Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
A mixture of erythrocytes of Blood Group Antigen B and glycoprotein fraction isolated from the saliva of secretors with Blood Group Antigen B.
Protein A/G purification
RUO
Primary
This monoclonal antibody reacts with human blood group B. It shows specific staining of erythrocytes and vascular epithelium of blood group B controls and no staining in group A controls. This antibody is applicable for tissue staining in tumor patients with blood groups B and AB.