Learn More
GE Healthcare Blot Processing Trays
Processor Plus is a novel fluid delivery system that automates acrylamide gel staining and western blot processing
Manufacturer:  GE Healthcare 80644461
Catalog No. 45-002-204
Add to cart
Description
Description
Processor Plus is a novel fluid delivery system that automates acrylamide gel staining and western blot processing. Changing from one application to the other is achieved by a fast, simple switch of a tray. Select a programmed protocol and push “start”. Processor Plus will then fill and empty the blot or gel tray with the correct solution at the right time. The blot-processing capabilities of Processor Plus include trays for four mini (up to 9 x 9.5cm) or two standard (up to 16 x 16cm) sized blots at one time. Five preprogrammed and five user-defined blot protocols are incorporated for flexibility, and microprocessor precision ensures consistent results with low background from membrane to membrane. Alternatively, the automated gel staining capabilities of Processor Plus include nine preprogrammed and four user-defined protocols for proteins and nucleic acids. A choice of small and large-format tray sizes accommodates one to six mini-gels or one to two standard size gels for silver or Coomassie blue-staining. The components behind the intelligent fluid handling of Processor Plus include a high-speed peristaltic pump for fast delivery and removal of reagents; a 10-port valve for the creation of multiple reagent protocols and segregation of hazardous waste; and a rhomboid-shaped tray with smooth and even rocking for rapid fluid mixing and coverage. Protocol entry and editing is simple with up to 30 steps allowed per protocol. A removable protocol key, which stores one protocol, protects critical protocols from accidental alteration. A serial printer can be connected to the Processor Plus for report generation. The report provides information on each step including the volume of reagents used, the time (in minutes) for each step, and a validation report form, with spaces for sample ID, date, and operator.
  • One system for gel staining and blot processing.
  • Automated staining of Coomassie blue or silver stain gels for proteins and nucleic acid analysis.
  • Prepares blots for high-sensitivity ECL and chromogenic detection.
  • Programmable control of protocol, solution, volume, and processing time.
  • Proven design for consistent, high-quality performance every time.
Specifications
Specifications
Automates Acrylamide Gel Staining and Western Blot Processing
16 x 16 cm
2 Pack
Blot Processing Trays