Learn More
BLOTTO antifoam in PBS, with 0.02% sodium azide, Thermo Scientific™
Blocking reagent for immunohistochemistry experiments
$100.90
Specifications
Product Type | Blocking Buffer |
---|---|
Form | Liquid |
For Use With (Application) | Immunohistochemistry (IHC) |
Content And Storage | -30°C to -10°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ61888AP
|
Thermo Scientific Chemicals
J61888AP |
500 mL |
Each of 1 for $100.90
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Blocking Buffer | |
Liquid | |
Immunohistochemistry (IHC) | |
-30°C to -10°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.