Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ BLOTTO in PBS, with 0.02% sodium azide

Blocks non-specific binding site for antibodies on the membrane

$39.80 - $65.90

Specifications

Storage Requirements -30°C to -10°C
Format Liquid
For Use With (Application) Blocks non-specific binding site for antibodies on the membrane
Product Type Blocking Buffer
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ60166AP View Documents Alfa Aesar™
J60166AP
500 mL Each for $65.90
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ60166AK View Documents Alfa Aesar™
J60166AK
250 mL Each for $39.80
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

-30°C to -10°C
Liquid
Blocks non-specific binding site for antibodies on the membrane
Blocking Buffer
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit