Learn More
Azzota Corp BLU-285 DFS Item
Manufacturer:  Azzota Corp C1137
BLU-285 is a potent and selective inhibitor of PDGFRα(D842V) and KIT Exon 17 mutants. Please request if need more quantities
Catalog No. 50-161-3105