Learn More
Selleck Chemical LLCSupplier Diversity Partner BLU9931 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S78195MG
BLU9931 is a potent, selective, and irreversible FGFR4 inhibitor with IC50 of 3 nM, about 297-, 184-, and 50-fold selectivity over FGFR1/2/3, respectively.
Catalog No. 50-194-8011