Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Sklar Blue Bozeman Forceps, Sklar 10-1/2 in., Straight, Serrated, Stainless Steel with Non-Conductive Nylon Coating, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 915307
Forceps, Sklar Blue Bozeman Forceps, Sklar, 10-1/2 in., Straight, Serrated, Stainless Steel with Non-Conductive Nylon Coating, EA. Sklar Blue Bozeman Forceps are a multi-functional instrument commonly used in gynecological procedures. They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity. The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. The jaws are available either curved or straight. Sklar Blue electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue instruments are fully autoclavable and may also be cleaned ultrasonically. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-1869