Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Screw Cap, 2 mL for Lysing Matrix tubes

Used in FastPrep systems to hold sample for homogenization.

Manufacturer:  MP Biomedicals™ 115065005

Catalog No. 11-506-5005


Add to cart

Description

Description

Applications: Blue Caps fit EMPTY FASTPREP tubes and are used in Fastprep systems to hold sample for homogenization. Available separately for those who wish to fill their own Lysing Matrix Tubes.

Product Description: Lysing Matrix B tubes have blue caps found in Fast RNA Pro Blue Kit which are used for isolation of total RNA from gram positive and gram negative bacteria.

Recommended Use: Use with Empty FastPrep Tubes. Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer and apply cap. Process using FastPrep instrument or other bead-beater to lyse, grind or homogenize sample.

Store at Room Temperature (15 to 30°C).

Specifications

Specifications

2 mL Screw Cap with O-ring for 2 ml microfuge tube (Lysing Matrix Tube)
1000
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.