Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMAL1 Mouse anti-Human, Biotin, Clone: OTI1C11, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270207B
Description
BMAL1 Monoclonal antibody specifically detects BMAL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
BMAL1 | |
OTI1C11 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Affinity Purified, Conjugated | |
ARNTL | |
Full length recombinant protein of human BMAL1 (NP_001169) produced in HEK293T cells. | |
Immunogen affinity purified | |
Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Hypoxia, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators, Vision | |
Primary | |
406.0 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Biotin | |
PBS with 0.05% Sodium Azide | |
aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title