Learn More
BMI-1 Mouse anti-Human, Mouse, Clone: AF27, Active Motif™
Mouse Monoclonal Antibody
Manufacturer:  Active MotifSupplier Diversity Partner 39993
Encompass
 View more versions of this product
Catalog No. 50-199-3023
Add to cart
Description
Description
Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin-protein ligase activity of RNF2/RING2.
Specifications
Specifications
BMI-1
ChIP-sequencing, Western Blot
1 mg/mL
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
P25916, P35226
AW546694; B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; Bmi1; Bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; BMI1 proto-oncogene, polycomb ring finger; FLVI2/BMI1; flvi-2/bmi-1; murine leukemia viral (bmi-1) oncogene homolog; PCGF4; Pcgf4 antibody; Polycomb complex protein BMI-1; polycomb group protein Bmi1; polycomb group ring finger 4; Polycomb group RING finger protein 4; RING finger protein 51; RNF51; RP11-573G6.1
This BMI-1 antibody was raised against a recombinant protein corresponding to full-length mouse BMI-1.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
BMI1
AF27
Unconjugated
BMI1
Liquid
Mouse
IgG2a
100 μg
-20°C, Avoid Freeze/Thaw Cycles
Primary
12151, 648