Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMI-1 Monoclonal Antibody (AF27), Active Motif™

Mouse Monoclonal Antibody
Supplier: Active Motif 39994

Description
Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin-protein ligase activity of RNF2/RING2.Specifications
BMI-1 | |
Monoclonal | |
1 mg/mL | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
P25916, P35226 | |
BMI1 | |
This BMI-1 antibody was raised against a recombinant protein corresponding to full-length mouse BMI-1. | |
10 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
ChIP-sequencing, Western Blot | |
AF27 | |
Unconjugated | |
BMI1 | |
AW546694; B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; Bmi1; Bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; BMI1 proto-oncogene, polycomb ring finger; FLVI2/BMI1; flvi-2/bmi-1; murine leukemia viral (bmi-1) oncogene homolog; PCGF4; Pcgf4 antibody; Polycomb complex protein BMI-1; polycomb group protein Bmi1; polycomb group ring finger 4; Polycomb group RING finger protein 4; RING finger protein 51; RNF51; RP11-573G6.1 | |
Mouse | |
Protein G | |
RUO | |
12151, 648 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title