Learn More
BML-190, Tocris
Potent, selective CB2 ligand
Manufacturer:  R&D Systems 1383/10
Quantity 10mg
 View more versions of this product
Catalog No. 13-831-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
426.9
BML-190
C23H23ClN2O4
1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1-[2-(4-morpholinyl)-2-oxoethyl]-1H-indole
Specifications
10mg
>99%
426.9
BML-190
2854-32-2
C23H23ClN2O4
1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1-[2-(4-morpholinyl)-2-oxoethyl]-1H-indole
Safety and Handling
Recommended Storage : Store at Room Temperature