Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-1/PCP Mouse anti-Human, DyLight™ 755, Clone: OTI3E9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270255IR

Catalog No. NBP270255IR


Add to cart

Description

Description

BMP-1/PCP Monoclonal antibody specifically detects BMP-1/PCP in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

BMP-1/PCP
OTI3E9
Western Blot, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
BMP1
Recombinant protein fragment corresponding to amino acids 225-617 of human BMP-1/PCP (NP_001190) produced in E.coli.
Immunogen affinity purified
Cytokine Research
Primary
649.0
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
BMP-1, bone morphogenetic protein 1, EC 3.4.24, EC 3.4.24.19, FLJ44432, Mammalian tolloid protein, mTld, PCOLC, PCP, PCP2, procollagen C-endopeptidase, Procollagen C-proteinase, TLD
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit